Would you like to log in again?
PFKFB1
Supplier: Rockland Immunochemicals
009-001-S87 009-001-S87S
75930-836EA 714 USD InStock
75930-836 75930-834
PFKFB1
Proteins and Peptides
PFKFB1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PFKFB1 is a member of the family of bifunctional 6-phosphofructo-2-kinase: fructose-2, 6-biphosphatase enzymes which forms a homodimer that catalyzes both the synthesis and degradation of fructose-2,6-biphosphate using independent catalytic domains (1). Fructose-2,6-biphosphate is an activator of the glycolysis pathway and an inhibitor of the gluconeogenesis pathway. Regulation of fructose-2,6-biphosphate levels through the activity of this enzyme is thought to regulate glucose homeostasis (2). PFKFB1 Protein is ideal for investigators involved in Signaling Proteins, Fructose Kinase Proteins, Cancer, Cell Cycle, and Metabolic Disorder research.
PFKFB1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PFKFB1 Protein is suitable for use in Western Blot. Expect a band approximately ~80kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty