Would you like to log in again?
PFKFB3
Supplier: Rockland Immunochemicals
009-001-S88 009-001-S88S
75930-840EA 714 USD InStock
75930-840 75930-838
PFKFB3
Proteins and Peptides
PFKFB3 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PFKFB3 or 6-phosphofructo-2-kinase/fructose-2, 6-biphosphatase 3 is a potent activator of a key regulatory enzyme in the glycolysis pathway called phosphofructokinase. Phosphofructokinase regulates the steady-state concentration of fructose-2, 6-bisphosphate, a potent activator of key regulatory enzyme of the glycolysis pathway. PFKFB3 is bifunctional, although the phosphatase activity is low due to a mutation in an active site which can be upregulated after phosphorylation by protein kinases (1). PFKFB3 may be responsible for sustaining the high glycolytic flux of rapidly proliferating leukemia cells and may represent a molecular target for chemotherapeutic intervention (2). PFKFB3 Protein is ideal for investigators involved in Signaling Proteins, Fructose Kinase Proteins, Cancer, Cell Cycle, and Metabolic Disorder research.
PFKFB3 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PFKFB3 Protein is suitable for use in Western Blot. Expect a band approximately ~83kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty