Would you like to log in again?
PIK3R1
Supplier: Rockland Immunochemicals
009-001-S94S 009-001-S94
75930-920EA 704 USD InStock
75930-920 75930-918
PIK3R1
Proteins and Peptides
PI3K (p85α alpha) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
The PI3K comprises of a 110 kDa catalytic subunit and an 85 kDa regulatory subunit. A number of isoforms of the 110 kDa catalytic subunit and the 85 kDa regulatory subunit exist in cells. p85α modulates the interaction between PI3K and platelet-derived growth factor receptor (1). Furthermore, estrogen receptor isoform ER-alpha binds in a ligand-dependent manner to the p85-alpha regulatory subunit of PI3K. Stimulation with estrogen increases ER-alpha-associated PI3K activity, leading to the activation of protein kinase B/AKT and endothelial nitric oxide synthase (eNOS) (2). PI3K (p85 alpha) Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Invasion/Metastasis, Metabolic Disorder, Neurobiology, NfkB Pathway, and WNT Signaling research.
PI3K (p85 alpha) Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol. PI3K (p85 alpha) Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~86kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty