Would you like to log in again?
PI2KA
Supplier: Rockland Immunochemicals
009-001-S96 009-001-S96S
75930-928EA 714 USD InStock
75930-928 75930-926
PI2KA
Proteins and Peptides
PI4KA control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PI4KA encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4, 5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by PI4KA gene is a type III enzyme that is not inhibited by adenosine. PI4KA is highly expressed in placenta and brain (1). PI4KA is associated with the membranes of Golgi vesicles and vacuoles (2). PI4KA Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Invasion/Metastasis, Lipid Kinases, Metabolic Disorder, Neurobiology, NfkB Pathway, and WNT Signaling research.
PI4KA Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PI4KA Protein is suitable for use in Western Blot. Expect a band approximately ~120kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty