Would you like to log in again?
PRPF19
Supplier: Rockland Immunochemicals
009-001-T06S 009-001-T06
75931-346EA 714 USD InStock
75931-346 75931-344
PRPF19
Proteins and Peptides
PRPF19 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PRPF19 or pre-mRNA-processing factor 19 is the human homolog of yeast Pso4. PRPF19 is a gene essential for cell survival, DNA repair and association with terminal deoxynucleotidyl transferase (1). PRPF19 forms a stable complex with SETMAR (Metnase) which is a SET and transposase fusion protein in humans and plays a positive role in double-strand break (DSB) repair. PRPF19 mediates association of SETMAR with sites of DNA damage (2). SNEV is an orthologue of PRPF19 and is an evolutionarily conserved splicing factor whose oligomerization is necessary for spliceosome assembly. PRPF19 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Apoptosis/Autophagy, and Cell Cycle Proteins research.
PRPF19 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PRPF19 Protein is suitable for use in Western Blot. Expect a band approximately ~74kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty