Would you like to log in again?
RAS protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T15S 009-001-T15
75935-142EA 364 USD InStock
75935-142 75935-144
RAS protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
RAS control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
The RAS gene superfamily encodes a group of closely related 21,000 dalton (p21) proteins with special affinity for guanine nucleotides (GTP). RAS and several other cellular proteins with similar biochemical properties are collectively known as G-proteins and they play key roles in a wide variety of cellular activities, including cell growth, differentiation, secretion, and protein trafficking (1). There are three forms of RAS gene in cells termed H-RAS, N-RAS, and K-RAS. RAS proteins play a direct causal role in human cancer and in other diseases. Mutant H-RAS, N-RAS, and K-RAS occur in varying frequencies in different tumor types (2). Other members of the RAS superfamily may also contribute to cancer. RAS Protein is ideal for investigators involved in Signaling Proteins, G-Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, ERK/MAPK Pathway, Invasion/Metastasis, and Neurobiology research.
RAS Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. RAS Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~47kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty