Would you like to log in again?
RELB
Supplier: Rockland Immunochemicals
009-001-T18S 009-001-T18
75931-504EA 364 USD InStock
75931-504 75931-506
RELB
Proteins and Peptides
Rel B control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Rel B is part of the NFkB complex and forms heterodimeric complexes with p50 (NFKB1) and p52 (NFKB2) (1). The homodimeric complexes of Rel B alone do not show DNA-binding activity. IHC analysis has shown that Rel B expression correlated with dendritic cell activation. NFkB-inducing kinase NIK is required for osteoclastogenesis in response to pathologic stimuli and overexpression of Rel B rescues differentiation of mouse NIK -/- osteoclast precursors. Rel B is required for RANKL-induced osteoclastogenesis in vitro and for TNF -induced bone resorption in vivo. This indicates that the alternative NFkB pathway, via Rel B, plays an essential and unique role in RANKL signaling toward osteoclast development (2). Rel B Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, Inflammation, Neurobiology, and NFkB Pathway research.
Rel B Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. Rel B Protein is suitable for use in Western Blot. Expect a band approximately ~68kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty