Would you like to log in again?
RHEB protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-T20 009-001-T20S
75931-580EA 364 USD InStock
75931-580 75931-582
RHEB protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
RHEB control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
RHEB is a member of the small GTPase superfamily and encodes a lipid-anchored, cell membrane protein with five repeats of the RAS-related GTP-binding region. RHEB is vital in the regulation of growth and cell cycle progression. RHEB has GTPase activity and shuttles between a GDP-bound form and a GTP-bound form. Drosophila RHEB is a direct target of Tsc2 GTPase-activating protein (GAP) activity both in vivo and in vitro. Point mutations in the Drosophila RHEB gene inhibit cell growth while overexpression promotes cell growth. RHEB functions downstream of Tsc1 and Tsc2 in the mTOR signaling pathway. RHEB Protein is ideal for investigators involved in Signaling Proteins, G-Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, ERK/MAPK Pathway, Invasion/Metastasis, and Neurobiology research.
RHEB Protein is stored in 50mM NaPhosphate pH7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.2mM DTT, 25% glycerol. RHEB Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~21kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty