Would you like to log in again?
SIRT2
Supplier: Rockland Immunochemicals
009-001-T32S 009-001-T32
75931-914EA 364 USD InStock
75931-914 75931-916
SIRT2
Proteins and Peptides
SIRT2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SIRT2 is a member of the sirtuin family of proteins which are homologs to the yeast Sir2 protein. Sirtuin family contain a sirtuin core domain and are grouped into four classes with SIRT2 being a member of class I. Inhibition of SIRT2 can lead to neuroprotection in cellular and invertebrate models of Huntington's disease (1). Huntington's disease is characterized by increased sterols synthesis in neuronal cells and this process is reversed by SIRT2 inhibition. SIRT2 can deacetylate lys40 of alpha-tubulin both in vitro and in vivo (2). Knockdown of SIRT2 via small interfering RNA results in tubulin hyperacetylation. SIRT2 Protein is ideal for investigators involved in Signaling Proteins, Deacetylase/Demethylase Proteins, Cancer, Cell Cycle, and Inflammation research.
SIRT2 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SIRT2 Protein is suitable for use in Western Blot. Expect a band approximately ~64kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty