Would you like to log in again?
SMAD3
Supplier: Rockland Immunochemicals
009-001-T39 009-001-T39S
75932-038EA 714 USD InStock
75932-038 75932-036
SMAD3
Proteins and Peptides
SMAD3 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SMAD3 is a direct mediator of transcriptional activation by the TGFβ receptor. The activity of SMAD3 is regulated by the TGFβ receptors, and SMAD3 is phosphorylated and associated with the ligand-bound receptor complex. TGFβ stimulation leads to phosphorylation and activation of SMAD3, which form a complex with SMAD4 that accumulate in the nucleus and regulate transcription of target genes such as CDK inhibitor (1). SMAD3 containing a C-terminal truncation acts as a dominant-negative inhibitor of the normal TGFβ response. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2 (2). SMAD3 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, AKT/PKB Pathway, Angiogenesis, Cancer, Cell Cycle, Cellular Stress, ERK/MAPK Pathway, Inflammation, JAK/STAT Pathway, JNK/SAPK Pathway, NfkB Pathway, and WNT Signaling research.
SMAD3 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. SMAD3 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~77kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty