Would you like to log in again?
SNCA
Supplier: Rockland Immunochemicals
009-001-T45S 009-001-T45
75932-070EA 210 USD InStock
75932-070 75932-072
SNCA
Proteins and Peptides
SNCA control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SNCA is a member of the synuclein family which also includes beta- and gamma-synuclein. These proteins are abundantly expressed in the brain and alpha- and beta-synuclein selectively inhibit phospholipase D2 which is a highly conserved protein that is also abundant in neurons, especially presynaptic terminals (1). SNCA gene is not a major risk factor in familial Parkinson disease (2). SNCA may serve to integrate pre-synaptic signaling and membrane trafficking and SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimer's disease. SNCA Protein is ideal for investigators involved in Signaling Proteins, Microtubule/Actin Associated Proteins, Cancer, and Neurobiology research.
SNCA Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. SNCA Protein is suitable for use in Western Blot. Expect a band approximately ~18kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty