Would you like to log in again?
SNCB
Supplier: Rockland Immunochemicals
009-001-T46 009-001-T46S
75932-074EA 210 USD InStock
75932-074 75932-076
SNCB
Proteins and Peptides
SNCB control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SNCB is a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. SNCB is highly homologous to alpha-synuclein which is abundantly expressed in the brain and putatively inhibits phospholipase D2 selectively. SNCB may play a role in neuronal plasticity, is abundant in neurofibrillary lesions of patients with Alzheimer disease. SNCB is shown to be highly expressed in the substantia nigra of the brain, a region of neuronal degeneration in patients with Parkinson disease but no direct relation to Parkinson disease has been established (1). An alteration in SNCB may impair its normal inhibitory action on the formation of toxic alpha-synuclein fibrils, thereby indirectly contributing to disease pathogenesis (2). SNCB Protein is ideal for investigators involved in Signaling Proteins, Microtubule/Actin Associated Proteins, Cancer, and Neurobiology research.
SNCB Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. SNCB Protein is suitable for use in Western Blot. Expect a band approximately ~19kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty