Would you like to log in again?
SNCG
Supplier: Rockland Immunochemicals
009-001-T47S 009-001-T47
75932-080EA 313 USD InStock
75932-080 75932-078
SNCG
Proteins and Peptides
SNCG control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SNCG or synuclein, gamma (breast cancer-specific protein 1) is a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. SNCG gene may also be overexpressed in ovarian cancers (1). The overexpression of SNCG may indicate breast cancer malignant progression from benign or in situ carcinoma. SNCG may also play a role in motor neurons dysfunction and cell death (2). Synuclein proteins play an important role in regulating neurotransmitter release from specific populations of midbrain dopamine neurons through mechanisms that differ from those reported in other neurons. SNCG Protein is ideal for investigators involved in Signaling Proteins, Microtubule/Actin Associated Proteins, Cancer, and Neurobiology research.
SNCG Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. SNCG Protein is suitable for use in Western Blot. Expect a band approximately ~17.5kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty