Would you like to log in again?
STAT1
Supplier: Rockland Immunochemicals
009-001-T51S 009-001-T51
75935-392EA 364 USD InStock
75935-392 75935-394
STAT1
Proteins and Peptides
STAT1 beta control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
STAT1β is a member of the signal transducers and activators of transcription (STAT) family of proteins that carry out a dual function: signal transduction and activation of transcription. STAT1β transcription factor is specific for the IFN pathway and plays a central role in mediating many, if not all, IFN-dependent biological responses (1). Presence of STAT1β leads to an efficient antiviral response when cells were infected with virus suggesting that a STAT-dependent pathway is activated following virus infection by endogenously produced IFN. Virus-induced STAT protein translocation from the cytoplasmic compartment can be detected within 3 h of infection. STAT1β Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, Inflammation, and JAK/STAT Pathway research.
STAT1 beta Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. STAT1 beta Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~118kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty