Would you like to log in again?
STAT4
Supplier: Rockland Immunochemicals
009-001-T55S 009-001-T55
75932-204EA 364 USD InStock
75932-204 75932-206
STAT4
Proteins and Peptides
STAT4 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
STAT4 (signal transducer and activator of transcription 4) is a member of the STAT family that is phosphorylated by receptor-associated protein kinases. STATs form homo- or heterodimers and translocate to the cell nucleus where they act as transcription activators. T helper (Th) cells differentiation is regulated by STAT4 (1). STAT4 controls T1/T2 balance during the response to live influenza virus and isolated viral proteins. The differentiation of gamma interferon (IFN-gamma)-producing Th1 and Tc1 cells after inoculation of live virus occurred independently of STAT4 expression. STAT4 has been shown to control pathogenesis of chronic obstructive pulmonary disease (2). STAT4 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, Inflammation, and JAK/STAT Pathway research.
STAT4 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. STAT4 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~87kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty