Would you like to log in again?
STAT5
Supplier: Rockland Immunochemicals
009-001-T56S 009-001-T56
75932-212EA 714 USD InStock
75932-212 75932-210
STAT5
Proteins and Peptides
STAT5 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
STAT5 (signal transducers and activators of transcription 5) is a member of the STAT family and this protein is phosphorylated by receptor associated protein kinases in response to cytokines and growth factors (1). The differentiation of T helper (Th) cells is regulated by members of the STAT family of signaling molecules. In BCR-ABL-positive cells, STAT5 is constitutively activated by tyrosine phosphorylation. STAT5 activation results in upregulation of Bcl-XL and increased resistance to induction of apoptosis. STAT5 is involved in expression and growth hormone-mediated sexually dimorphic regulation of cytochrome P450 3A10/lithocholic acid 6 beta-hydroxylase (2). STAT5 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Apoptosis/Autophagy, Cancer, ERK/MAPK Pathway, Inflammation, and JAK/STAT Pathway research.
STAT5 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. STAT5 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~92kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty