Would you like to log in again?
STUB1
Supplier: Rockland Immunochemicals
009-001-T64S 009-001-T64
75932-364EA 313 USD InStock
75932-364 75932-362
STUB1
Proteins and Peptides
STUB1 (CHIP) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
CHIP (STUB1) or STIP1 homology and U-box containing protein 1 (also known as E3 ubiquitin protein ligase) is an ubiquitin ligase/cochaperone that participates in protein quality control by targeting a broad range of chaperone protein substrates for degradation (1). CHIP acts as a molecular switch for the degradation of misfolded proteins via the proteasome and lysosome pathways (2). CHIP controls the cellular level of BER enzymes and, correspondingly, the efficiency and capacity of BER. CHIP E3 ubiquitin ligases act sequentially in ER membrane and cytosol to monitor the folding status of CFTR and delF508.CHIP possesses intrinsic E3 ubiquitin ligase activity and promotes ubiquitylation. STUB1 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cell Cycle, Cellular Stress, Cytoplasmic Tyrosine Kinases, and Metabolic Disorder research.
STUB1 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. STUB1 Protein is suitable for use in Western Blot. Expect a band approximately ~37kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty