Would you like to log in again?
SUMO1
Supplier: Rockland Immunochemicals
009-001-T66S 009-001-T66
75935-528EA 313 USD InStock
75935-528 75932-374
SUMO1
Proteins and Peptides
SUMO1 (UBL1) control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
SUMO1 (also known as UBL1) is a member of the SUMO (small ubiquitin-like modifier) protein family which functions in a manner similar to ubiquitin. SUMO1 is bound to target proteins as part of a post-translational modification system and it is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability (1). SUMO1 may be involved in the repair of TOP1-mediated DNA damage (2). The sumoylation pathway plays a significant role in mammalian DNA damage response. SUMO1 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Apoptosis/Autophagy, Cell Cycle, and NfkB Pathway research.
SUMO1 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. SUMO1 Protein is suitable for use in Western Blot. Expect a band approximately ~19kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty