Would you like to log in again?
Tau-441 (151-421)
Supplier: Rockland Immunochemicals
009-001-T87S 009-001-T87
75932-654EA 513 USD InStock
75932-654 75932-652
Tau-441 (151-421)
Proteins and Peptides
Tau-441 (151-421) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Tau-441 or Tau-F is a member of the Tau family of proteins which function to stabilize the microtubules by binding to them. Tau proteins are subject to phosphorylation and this phenomenon regulates the association of the Tau protein with the microtubules (1). Deposits of Alzheimer's disease AD-associated proteins, such as hyperphosphorylated Tau, as well as other shared misfolded proteins, such as, β-amyloid precursor protein (βAPP), ubiquitin, and various chaperones and protein kinases are thought to play a pathologic role in the cognitive decline and muscular failure. Malfunctioning of Tau proteins is associated with microtubules disintegration and collapsing of the neuronal transport system (2). Tau-441 Protein is ideal for investigators involved in Signaling Proteins, Tau Proteins, Invasion/Metastasis, Neurobiology, and p38 Pathway research.
Tau-441 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. Tau-441 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~32kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty