Would you like to log in again?
TRIM37 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U21S 009-001-U21
75933-124EA 714 USD InStock
75933-124 75933-122
TRIM37 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
TRIM37 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TRIM37 or tripartite motif containing 37 is a member of the tripartite motif (TRIM) family whose members are involved in diverse cellular functions such as developmental patterning and oncogenesis (1). The TRIM motif includes zinc-binding domains, a RING finger region, a B-box motif and a coiled-coil domain. The RING finger and B-box domains chelate zinc and might be involved in protein-protein and/or protein-nucleic acid interactions. Mutations in TRIM37 are associated with mulibrey (muscle-liver-brain-eye) nanism, an autosomal recessive disorder that involves several tissues of mesodermal origin (2). TRIM37 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cell Cycle, Cellular Stress, Inflammation, Invasion/Metastasis, and Neurobiology research.
TRIM37 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TRIM37 Protein is suitable for use in Western Blot. Expect a band approximately ~140kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty