Would you like to log in again?
TSSK3 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-U23 009-001-U23S
75935-472EA 364 USD InStock
75935-472 75935-474
TSSK3 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
TSSK3 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TSSK3 is a serine/threonine kinase belonging to the subfamily of testis-specific kinases 3 which expressed exclusively in the testis that is thought to play a main role in either germ cell differentiation or mature sperm function (1). TSSK3 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cardiovascular Disease, and Ser/Thr Kinases research.
TSSK3 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TSSK3 Protein is suitable for use in Western Blot. Expect a band approximately ~56kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty