Would you like to log in again?
UBA3
Supplier: Rockland Immunochemicals
009-001-U26 009-001-U26S
75933-300EA 714 USD InStock
75933-300 75935-482
UBA3
Proteins and Peptides
UBA3(UBE1C), control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UBA3 (also known as UBE1C) or ubiquitin-like modifier activating enzyme 3 is a member of the E1 ubiquitin-activating enzyme family. UBA3 associates with AppBp1, an amyloid beta precursor protein binding protein, to form a heterodimer, and then the enzyme complex activates NEDD8, a ubiquitin-like protein, which regulates cell division, signaling and embryogenesis. The mechanism of NEDD8 activation by APPBP1-UBA3 shows a high degree of conservation with ubiquitin activation by UBA1 (1). The NEDD8-activating enzyme, or NAE, composed of NAE1 and UBA3 subunits, is an essential component for the treatment of cancer disease (2). UBA3 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Cancer, Cell Cycle, and Neurobiology research.
UBA3 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. UBA3 Protein is suitable for use in Western Blot. Expect a band approximately ~73kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty