Would you like to log in again?
UHMK1
Supplier: Rockland Immunochemicals
009-001-U53 009-001-U53S
75933-422EA 364 USD InStock
75933-422 75933-424
UHMK1
Proteins and Peptides
UHMK1 (KIS) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
UHMK1 (KIS) or U2AF homology motif (UHM) kinase 1 is a serine/threonine protein kinase that promotes cell cycle progression through G1 phase. UHMK1 phosphorylates the cyclin-dependent kinase inhibitor 1B (p27Kip1) and this results in its export to the nucleus where it undergoes degradation. UHMK1 is a growth factor-dependent nuclear kinase which can regulate cell cycle progression (1). Elevated levels of UHMK1 protein in leukemia cells has been shown to promote cell cycle progression. UHMK1 has also been shown to function in the adult nervous system where it is highly expressed and the gene has been associated with schizophrenia (2). UHMK1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, Cell Cycle, and Ser/Thr Kinases research.
UHMK1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. UHMK1 Protein is suitable for use in Western Blot. Expect a band approximately ~65kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty