Would you like to log in again?
Human Recombinant Artemin
Supplier: Rockland Immunochemicals
009-001-U64-0100 009-001-U64-1000 009-001-U64-0020 009-001-U64-0005
10800-090EA 704 USD InStock
10800-090 10800-092 10800-094 10800-088
Human Recombinant Artemin
Proteins and Peptides
Recombinant Human Artemin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Artemin is a novel member of the glial cell line-derived neurotrophic factor (GNDF) ligand family. Current evidence suggests that Artemin signals through the receptor complex, GFRα3-RET, to influence neuron survival in vitro and in vivo. Recombinant human Artemin is a non-glycosylated, disulfide-linked homodimer, containing two 113 amino acid chains, with a total molecular weight of 24.2 kDa.
Recombinant human Artemin is suitable as a control for polyclonal or monoclonal anti-Artemin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty