Would you like to log in again?
Human Recombinant Chemerin
Supplier: Rockland Immunochemicals
009-001-U70-0005 009-001-U70-0020 009-001-U70-1000 009-001-U70-0100
10800-786EA 704 USD InStock
10800-786 10800-788 10800-790 10800-784
Human Recombinant Chemerin
Proteins and Peptides
Recombinant Human Chemerin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Chemerin is a chemoattractant expressed in white adipose, liver and lung tissues. Chemerin is a ligand for the G-protein coupled receptor known as ChemR23 (or chemokine-like receptor-1), which is expressed mainly on dendritic cells, macrophages and some adipocytes. Recombinant human Chemerin is a non-glycosylated protein, containing 138 amino acids, with a total molecular weight of 16 kDa.
Chemerin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Chemerin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty