Would you like to log in again?
Human Recombinant FGF-22
Supplier: Rockland Immunochemicals
009-001-U78-1000 009-001-U78-0005 009-001-U78-0100 009-001-U78-0020
10802-868EA 140 USD InStock
10802-868 10802-864 10802-866 10802-862
Human Recombinant FGF-22
Proteins and Peptides
Recombinant Human FGF-22 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fibroblast Growth Factors (FGFs) are a 22 member family of proteins known to be involved in angiogenesis, wound healing and embryonic development. As a family, they bind to heparin and signal through four receptor tyrosine kinases called, FGFR1, 2, 3 and 4. Human FGF-22 is a member of the FGF-7 subfamily and is synthesized by multiple cell lines including neurons, keratinocytes and skeletal muscle myotubes. Human FGF-22 shares 86% homology with mouse FGF-22. Recombinant human FGF-22 is a non-glycosylated protein, containing149 amino acids, with a molecular weight of 17.3 kDa.
Fibroblast Growth Factor-22 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor-22 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty