Would you like to log in again?
Human Recombinant Glial Derived NTF
Supplier: Rockland Immunochemicals
009-001-V10-0100 009-001-V10-1000 009-001-V10-0002 009-001-V10-0010
10801-708EA 326 USD InStock
10801-708 10801-714 10801-702 10801-720
Human Recombinant Glial Derived NTF
Proteins and Peptides
Recombinant Human Glial Derived NTF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Glial Cell Line-Derived Neurotrophic Factor (GDNF) is a neurotrophic factor that is closely related to other neurotrophic factors, such as Neurturin, Persephin, and Artemin, by a common structural feature called the cysteine-knot. GDNF signals through a multicomponent system of receptors that includes RET and GFRα1-4, to promote dopamine uptake, survival and differentiation of neurons. Recombinant human GDNF is a non-glycosylated homodimer, containing two 135 amino acid chains, with a total molecular weight of 30.4 kDa.
Glial Derived Neurotrophic Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Glial Derived Neurotrophic Factor in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty