Would you like to log in again?
Human Recombinant INF gamma
Supplier: Rockland Immunochemicals
009-001-V20-0020 009-001-V20-0100 009-001-V20-1000
75928-986EA 326 USD InStock
75928-986 75928-984 75928-988
Human Recombinant INF gamma
Proteins and Peptides
Recombinant Human INF gamma control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interferon gamma, IFNγ, is a major immune-modulating molecule produced mainly by T cells and natural killer cells activated by antigens, mitogens or alloantigens. Most immune cells express IFN-γ receptors and respond to IFNγ-induced signaling by up-regulating MHC class I expression. Human IFNγ does not show cross-reactivity with mouse. Recombinant human IFNγ is a non-glycosylated protein, containing 144 amino acids, with a molecular weight of 16.88 kDa.
Interferon-gamma Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interferon-gamma in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty