Would you like to log in again?
Human Recombinant IL-10
Supplier: Rockland Immunochemicals
009-001-V47-1000 009-001-V47-0010 009-001-V47-0002 009-001-V47-0100
75929-020EA 137.15 USD InStock
75929-020 75929-014 75929-016 75929-018
Human Recombinant IL-10
Proteins and Peptides
Recombinant Human IL-10 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 10 (IL-10) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including, IFN-γ, TNF-α, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Recombinant human IL-10 is a non-glycosylated protein, containing 161 amino acids, with a molecular weight of 16.6 kDa.
Human Interleukin-10 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Human Interleukin-10 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty