Would you like to log in again?
Human Recombinant IL-17E (IL-25)
Supplier: Rockland Immunochemicals
009-001-V57-0005 009-001-V57-1000 009-001-V57-0025 009-001-V57-0100
75934-702EA 704 USD InStock
75934-702 75934-710 75929-092 75934-694
Human Recombinant IL-17E (IL-25)
Proteins and Peptides
Recombinant Human IL-17E (IL-25) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-17E (IL-17E), also commonly called IL-25, is a pro-inflammatory cytokine member of a six-species family of proteins (IL-17A-17F). IL-17F stimulates secretion of IL-8 and induces activation of NF-kB by binding to the receptor IL-17RB. Recombinant human IL-17E is a non-glycosylated, disulfide-linked, homodimer, containing two 145 amino acid chains, with a total molecular weight of 35.5 kDa.
Interleukin-17E Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-17E in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty