Would you like to log in again?
Human Recombinant IL-20
Supplier: Rockland Immunochemicals
009-001-V60-1000 009-001-V60-0002 009-001-V60-0010 009-001-V60-0100
75934-738EA 137.15 USD InStock
75934-738 75934-736 75934-734 75934-732
Human Recombinant IL-20
Proteins and Peptides
Recombinant Human IL-20 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-20 (IL-20) is an IL-10 family member produced by keratinocytes and monocytes. IL-20 appears to regulate the growth of keratinocytes during inflammation of the skin. Recombinant human IL-20 is a non-glycosylated homodimer, containing two 153 amino acids, with a total molecular weight of 35.2 kDa.
Interleukin-20 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-20 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty