Would you like to log in again?
Human Recombinant CCL8
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
009-001-V75-0100 009-001-V75-0010
75930-040EA 326 USD InStock
75930-040 75930-042
Human Recombinant CCL8
Proteins and Peptides
Recombinant Human CCL8 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Monocyte Chemotactic Protein 2 (MCP-2), also known as CCL8, is implicated in allergic responses through its ability to activate mast cells, eosinophils, and basophils. MCP-2 is known to signal through several G protein-coupled receptors including, CCR1, CCR2B, and CCR5. Recombinant human MCP-2 is a non-glycosylated protein, containing 76 amino acids, with a molecular weight of 8.9 kDa.
Monocyte Chemotactic Protein-2 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Monocyte Chemotactic Protein-2 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty