Would you like to log in again?
Human Recombinant Myostatin
Supplier: Rockland Immunochemicals
009-001-W01-1000 009-001-W01-0002 009-001-W01-0100 009-001-W01-0010
75930-180EA 137.15 USD InStock
75930-180 75930-178 75930-176 75930-174
Human Recombinant Myostatin
Proteins and Peptides
Recombinant Human Myostatin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Myostatin is a member of the TGF-β superfamily that is essential for proper regulation of skeletal muscle mass in a number of species ranging from fish to humans. Myostatin is a secreted protein that negatively regulates skeletal muscle mass determining both muscle fiber number and size. Inhibition of myostatin can increase muscle mass in a number of animal models of human disease, including muscular dystrophy. Recombinant human Myostatin is a non-glycosylated homodimer, containing two 109 amino acid chains, with a total molecular weight of 24.8 kDa.
Myostatin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Myostatin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty