Would you like to log in again?
Human Recombinant PDGF-AA
Supplier: Rockland Immunochemicals
009-001-W08-0002 009-001-W08-0010 009-001-W08-1000 009-001-W08-0100
75931-090EA 137.15 USD InStock
75931-090 75931-088 75931-084 75931-086
Human Recombinant PDGF-AA
Proteins and Peptides
Recombinant Human PDGF-AA control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Platelet-Derived Growth Factor (PDGF) is a mitogenic peptide growth hormone carried in the alpha-granules of platelets and is released when platelets adhere to traumatized tissues. Connective tissue cells near the traumatized region respond by initiating the process of replication. The synthesis of PDGF can be induced by IL-1, IL-6, TNF-α, TGF-β and EGF. Recombinant human PDGF-AA is a non-glycosylated disulfide-linked homodimer, containing two 125 amino acid chains, with a total molecular weight of 28.5 kDa.
Platelet Derived Growth Factor-AA Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Platelet Derived Growth Factor-AA in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty