Would you like to log in again?
Human Prolactin Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-W15-0050 009-001-W15-1000 009-001-W15-0010
75931-182EA 2179 USD InStock
75931-182 75931-184 75931-186
Human Prolactin Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human Prolactin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Prolactin (PRL) is a hormone that is made and secreted by the pituitary gland and acts in an endocrine, paracrine and autocrine manner. The prolactin receptor is expressed on many cell types and is linked to JAK kinase signaling pathways. PRL’s primary function is to regulate lactation, but it is known to have many other regulation functions. These functions include roles in the immune system, cell growth, apoptosis, and differentiation. Recombinant human Prolactin is a non-glycosylated protein, containing 200 amino acids, with a molecular weight of 23 kDa.
Prolactin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Prolactin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty