Would you like to log in again?
Human recombinant resistin dimer (from E. coli)
Supplier: Rockland Immunochemicals
009-001-W19-1000 009-001-W19-0100 009-001-W19-0005 009-001-W19-0025
75931-538EA 1058 USD InStock
75931-538 75931-534 75931-546 75931-542
Human recombinant resistin dimer (from E. coli)
Proteins and Peptides
Recombinant Human Resistin Dimer control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Resistin is a peptide hormone belonging to the class of cysteine-rich secreted proteins which is termed the RELM family and is also described as ADSF (Adipose Tissue-Specific Secretory Factor) and FIZZ3 (Found in Inflammatory Zone). Resistin is produced by adipocytes and may be an important link between obesity and insulin resistance. Recombinant human Resistin is a disulfide-linked homodimer, containing two 93 amino acid chains , with a total molecular weight of 20 kDa.
Resistin Dimer Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Resistin Dimer in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty