Would you like to log in again?
Human TNF Receptor-1 Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-W29-0020 009-001-W29-0100 009-001-W29-1000 009-001-W29-0005
75933-224EA 4839 USD InStock
75933-224 75933-226 75933-228 75933-230
Human TNF Receptor-1 Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human TNF Receptor-1 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TNF Receptor 1 (TNFR1) is expressed in most tissues and is activated by soluble and membrane-bound TNFα. TNFR1 is known to activate NF-kB and MAPK pathways to induce inflammation, promote apoptotic cell death, inhibit tumorigenesis and inhibit viral replication. The soluble form of recombinant human TNFR1 is a non-glycosylated protein, containing 162 amino acids, with a molecular weight of 18.3 kDa.
Tumor Necrosis Factor Receptor Type 1 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Tumor Necrosis Factor Receptor Type 1 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty