Would you like to log in again?
Human TSLP Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-W35-0010 009-001-W35-0100 009-001-W35-1000 009-001-W35-0002
75932-896EA 137.15 USD InStock
75932-896 75932-894 75932-892 75932-890
Human TSLP Protein, Rockland Immunochemicals
Proteins and Peptides
Recombinant Human TSLP control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Thymic Stromal Lymphopoietin (TSLP) is a hematopoietic cytokine, produced in several tissues including heart, liver and prostate. TSLP sends signals through a heterodimeric receptor complex, containingthe TSLP-R and the IL-7Rα chain to induce the release of T cell-attracting chemokines from monocytes. Recombinant human TSLP is a non-glycosylated protein, containing 132 amino acids, with a molecular weight of 15 kDa.
Thymic Stromal Lymphopoietin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Thymic Stromal Lymphopoietin in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty