Would you like to log in again?
Human Recombinant VEGF-165 (from E. coli)
Supplier: Rockland Immunochemicals
009-F01-B99-0010 009-F01-B99-0002 009-F01-B99-0100 009-F01-B99-1000
75933-486EA 326 USD InStock
75933-486 75933-488 75933-490 75933-484
Human Recombinant VEGF-165 (from E. coli)
Proteins and Peptides
Recombinant Human VEGF-165 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Vascular Endothelial Growth Factor-A (VEGF-A) was originally isolated from tumor cells and is produced by a wide variety of cell types. In addition to stimulating vascular growth and vascular permeability, VEGF-A may play a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several variants, VEGF-165 being the most abundant. Recombinant human VEGF 165 is a non-glycosylated homodimer, containing two 165 amino acids, with a total molecular weight of 38.2 kDa.
Endothelial Growth Factor-165 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Endothelial Growth Factor-165 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty