Would you like to log in again?
Mouse IL-33
Supplier: Rockland Immunochemicals
010-001-B50-0010 010-001-B50-0002 010-001-B50-1000 010-001-B50-0100
75934-756EA 137.15 USD InStock
75934-756 75934-754 75934-752 75934-750
Mouse IL-33
Proteins and Peptides
Recombinant Mouse IL-33 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 33 (IL-33) is a member of the IL-1 cytokine family and is constitutively expressed in smooth muscle cells and the airway epithelium. IL-33 signals through receptors, IL-1RL1 and IL-1RAP, to induce type 2 cytokine production in polarized Th2 cells. Recombinant mouse IL-33 is a non-glycosylated protein, containing 159 amino acids, with a molecular weight of 17.7 kDa.
Interleukin-33 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-33 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty