Would you like to log in again?
Mouse IL-3
Supplier: Rockland Immunochemicals
010-001-B92-0002 010-001-B92-0010 010-001-B92-1000 010-001-B92-0100
75929-150EA 4582 USD InStock
75929-150 75929-152 75929-154 75934-740
Mouse IL-3
Proteins and Peptides
Recombinant Mouse IL-3 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-3 (IL-3) is a cytokine produced by activated T cells and mast cells. IL-3 is able to induce the differentiation of hematopoietic stem cells to precursor cells of myeloid lineage (erythrocytes, megakaryocytes, granulocytes, dendritic cells and monocytes). IL-3 also has functions in the nervous system and appears to be important in several chronic inflammatory diseases. Recombinant mouse IL-3 is a non-glycosylated protein, containing 135 amino acids, with a molecular weight of 15.1 kDa.
Human Interleukin-3 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Human Interleukin-3 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty