Would you like to log in again?
Mouse VEGF-165
Supplier: Rockland Immunochemicals
010-001-B99-1000 010-001-B99-0010 010-001-B99-0100 010-001-B99-0002
75933-500EA 728 USD InStock
75933-500 75933-504 75933-492 75933-496
Mouse VEGF-165
Proteins and Peptides
Recombinant Mouse VEGF-165 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Vascular Endothelial Growth Factor-A (VEGF-A) was originally isolated from tumor cells and is produced by a wide variety of cell types. In addition to stimulating vascular growth and vascular permeability, VEGF-A may play a role in stimulating vasodilation via nitric oxide-dependent pathways. VEGF-A has several variants, VEGF-165 being the most abundant. Rat and bovine VEGF are one amino acid shorter than the human factor, and the bovine and human sequences show a homology of 95%. Recombinant mouse VEGF-165 is a non-glycosylated, disulfide-linked homodimer, containing 165 amino acids, with a molecular weight of 39 kDa.
Endothelial Growth Factor-165 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Endothelial Growth Factor-165 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty