Would you like to log in again?
RANKL Mouse Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
010-001-C22
RL010-001-C22EA 329 USD InStock
RL010-001-C22
RANKL Mouse Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
Receptor Activator of Nuclear factor kappa-B Ligand Mouse Recombinant Protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
The secreted cytokine RANKL (Receptor Activator of Nuclear factor kappa-B Ligand) is critically involved in osteoclastic differentiation and activation and in the regulation of specific immunity. RANKL exists as a homotrimer, is glycosylated, and occurs in 3 forms: cell-bound RANKL, which is expressed by osteoblast lineage cells, soluble RANKL (sRANKL), which is expressed by activated T lymphocytes, and a truncated ectodomain form derived from the cell-bound RANK Ligand, which is enzymatically processed by TACE (TNF-alpha converting enzyme (TACE; ADAM-17)). All three forms stimulate their specific receptor, RANK, which is located on osteoclastic and dendritic cells. RANKL binds to TNFRSF11B/OPG and to TNFRSF11A/RANK. RANKL augments the ability of dendritic cells to stimulate naive T-cell proliferation. It may be an important regulator of interactions between T-cells and dendritic cells and may play a role in the regulation of the T-cell-dependent immune response. It may also play an important role in enhanced bone-resorption in humoral hypercalcemia of malignancy. Deficiency in Tnfsf11 results in failure to form lobulo-alveolar mammary structures during pregnancy, resulting in death of newborns. Trance-deficient mice show severe osteopetrosis.
RANKL is suitable as a control for polyclonal or monoclonal anti-RANKL in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty