Would you like to log in again?
Mouse Gro-alpha /KC /CXCL1
Supplier: Rockland Immunochemicals
010-001-V15-0005 010-001-V15-1000 010-001-V15-0020 010-001-V15-0100
75928-640EA 625 USD InStock
75928-640 75928-644 75928-642 75928-638
Mouse Gro-alpha /KC /CXCL1
Proteins and Peptides
Recombinant Mouse Gro-alpha /KC /CXCL1 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
GROα, also known as CXCL1, is a chemokine thought to have mitogenic properties and chemoattract neutrophils. Secreted by macrophages, epithelial cells, neutrophils and melanomas, GROα signals through chemokine receptor, CXCR2, and has been implicated in the processes of spinal cord formation, inflammation, angiogenesis, tumorigenesis, and wound healing. Recombinant mouse GROα is a non-glycosylated protein, containing 72 amino acids, with a molecular weight of 7.8 kDa.
Gro Alpha Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Gro Alpha in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty