Would you like to log in again?
Mouse INF gamma
Supplier: Rockland Immunochemicals
010-001-V20-0020 010-001-V20-0100 010-001-V20-1000
76221-882EA 1732 USD InStock
76221-882 75928-964 75928-968
Mouse INF gamma
Proteins and Peptides
Recombinant Mouse INF gamma control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interferon gamma, IFNγ, is a major immune-modulating molecule produced mainly by T cells and natural killer cells activated by antigens, mitogens or alloantigens. Most immune cells express IFNγ receptors and respond to IFNγ-induced signaling by up-regulating MHC class I expression. Human IFNγ does not show cross-reactivity with mouse. Recombinant mouse IFNγ is a non-glycosylated protein, containing 134 amino acids, with a molecular weight of 15.6 kDa.
Interferon-gamma Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interferon-gamma in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty