Would you like to log in again?
Mouse Recombinant IL-2
Supplier: Rockland Immunochemicals
010-001-V27-0020 010-001-V27-1000 010-001-V27-0100 010-001-V27-0005
75934-728EA 326 USD InStock
75934-728 75934-726 75934-724 75934-730
Mouse Recombinant IL-2
Proteins and Peptides
Recombinant Mouse IL-2 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-2 (IL-2) is an immuno-modulatory cytokine produced by stimulated T cells. IL-2 signals through the IL-2R receptor to induce proliferation of activated T cells. IL-2 also stimulates proliferation and differentiation of B cells, natural killer cells, monocytes and macrophages. Recombinant mouse IL-2 is a non-glycosylated protein, containing 149 amino acids, with a molecular weight of 17.2 kDa.
Human Interleukin-2 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Human Interleukin-2 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty