Would you like to log in again?
Mouse Recombinant IL-12
Supplier: Rockland Immunochemicals
010-001-V51-0002 010-001-V51-1000 010-001-V51-0100 010-001-V51-0010
75929-050EA 1932 USD InStock
75929-050 75929-052 75929-046 75929-048
Mouse Recombinant IL-12
Proteins and Peptides
Recombinant Mouse IL-12 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-12 (IL-12) is an important mediator of cellular-immunity produced by dendritic cells, macrophages and B cells when stimulated with antigen. IL-12 is involved in differentiating precursor helper T cells to Th1 and/or Th2 subtypes and promotes the production of IFN gamma and TNF alpha. Mouse IL-12 is active on human and mouse cells. Recombinant mouse IL-12 is a glycosylated, disulfide-linked heterodimer. It is containing one p35 su
Interleukin-12 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-12 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty