Would you like to log in again?
Mouse IL-16
Supplier: Rockland Immunochemicals
010-001-V55-0010 010-001-V55-0002 010-001-V55-1000 010-001-V55-0100
75928-784EA 924 USD InStock
75928-784 75928-776 75928-788 75928-778
Mouse IL-16
Proteins and Peptides
Recombinant Mouse IL-16 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 16 (IL-16) is produced primarily by CD4+ and CD8+ T cells and acts as a chemo-attractant for lymphocytes, monocytes, eosinophils, dendritic cells and Langerhans cells. Additionally, IL-16 has been reported to upregulate IL-2 receptor (CD25), induce progression of cells to the G1 phase and suppress HIV & SIV replication. Recombinant mouse IL-16 is a non-glycosylated protein, containing 127 amino acids, with a molecular weight of 13.2 kDa.
Interleukin-16 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-16 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty