Would you like to log in again?
Mouse Recombinant LIF
Supplier: Rockland Immunochemicals
010-001-V72-1000 010-001-V72-0100 010-001-V72-0005 010-001-V72-0020
75929-580EA 899 USD InStock
75929-580 75929-582 75934-848 75933-812
Mouse Recombinant LIF
Proteins and Peptides
Recombinant Mouse LIF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Mouse Leukemia Inhibitory Factor (LIF) is member of the IL-6 family and is made by a variety of tissues. LIF signals through receptors gp190 (LIFR) and gp130 to activate STAT3 which ultimately promotes mouse embryonic stem cell self-renewal and pluripotentcy of long term cultures. This activity is similar to that attributed to FGF-basic (FGF-2) for human embryonic stem cells. Recombinant mouse LIF is a non-glycosylated protein, containing 181 amino acids, with a molecular weight of 20 kDa.
Leukemia Inhibitory Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Leukemia Inhibitory Factor in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty