Would you like to log in again?
Mouse Recombinant IL-27 (p28) (from E. coli)
Supplier: Rockland Immunochemicals
010-F01-B54-1000 010-F01-B54-0100 010-F01-B54-0002
75928-806EA 1008 USD InStock
75928-806 75928-804 75928-808
Mouse Recombinant IL-27 (p28) (from E. coli)
Proteins and Peptides
Recombinant Mouse IL-27 (p28) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
The p28 subunit of IL-27 (IL-27/p28), also known as Interleukin-30 (IL-30), is a member of the IL-12 family of cytokines. When combined with EBI3 (Epstein-Barr virus induced gene 3), p28 forms a heterodimer known as IL-27. p28 is thought to be a pro-inflammatory cytokine inducing immuno-modulatory effects, but the details of these effects are currently unclear and research is focused on delineating its specific biological functions. Recombinant mouse p28 is a non-glycosylated protein, containing 207 amino acids, with a molecular weight of 23.7 kDa.
Interleukin-27 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-27 in immunological assays. Buffer Formulation: 10 mM sodium bicarbonate, pH 8.5.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty